missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Well Plate Accessories

Product Code. 15162779
Click to view available options
Product Type:
Automated Sample Concentrator
Center Hole Cap
Deep Well Plate Vial
MTRS Vial
Manual Concentrator
Mat Applicator
Microtiter Plate Vial
Plug
Sealing Tape Applicator
Square Well Plate Vial
Tube Tray
Volume (Metric) Working:
100 μL
1000 μL
1100 μL
1500 μL
350 μL
500 μL
700 μL
800 μL
Not Applicable
Well Shape:
Flat Bottom (F-well)
Not Applicable
Round
Square
Taper
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 2
Pack of 500
Pack of 96
Pack of 960
This item is not returnable. View return policy

Product Code. 15162779

Brand: Thermo Scientific™ 60180V105

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.

Sealing Tape Applicator
Ensure a consistent, reliable seal with all microplate formats and all types of tape with this easy to use, handheld applicator for Thermo Scientific™ WebSeal Sealing Tapes. They are packaged two per box.

Sample Concentrators
Remove most chromatography solvents in just minutes. Simple to operate and maintain, Thermo Scientific™ WebSeal Sample Concentrators allow quick, easy solvent evaporation from 96 well microplates, offering significantly quicker dry-down times than standard methods such as vacuum ovens or centrifuges. These evaporators offer significant throughput advantages for laboratories looking to accelerate microplate sample preparation. Depending on your throughput requirements, choose either the manually operated MiniVap Concentrator or the programmable, robot-ready UltraVap.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Crimp Top
Product Line Webseal
Color Clear
Product Type Deep Well Plate Vial
For Use With (Equipment) 96-well Square Deep-well Plates
Material Glass
No. of Wells 96
Volume (Metric) Working 1100 μL
Total Volume (Metric) Well 1100 μL
Well Shape Round
Plate Compatibility 96-well Plates
Length (Metric) 7.45 mm
Width (Metric) 45.0 mm
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.