missing translation for 'onlineSavingsMsg'
Learn More

DiversifiedBiotech Well Pads

Product Code. 30282300
Change view
Click to view available options
Product Type:
Microplate Well Pad
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30282300 Microplate Well Pad Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30282300 Supplier DiversifiedBiotech Supplier No. WPNP1000
  • ONLINE EXCLUSIVE PRICE
    £15.95
    £13.32 / Pack of 50
    Save £2.63
    16% Off
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fast and easy note-taking for microplate samples

  • A note pad that corresponds to the layout of a 96-well plate
  • A fast and easy way to keep accurate records of samples in a microplate
  • The adhesive backing allows the researcher to affix individual sheets into a notebook
  • The pad contains (50) 5 x 7 sheets
  • Fast and easy note-taking for microplate samples

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Color Black, Blue
Disposable Single-use
Product Type Microplate Well Pad
For Use With (Equipment) Well Plates
Material Plastic
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Square
For Use With (Application) Note-taking
Includes Well Pad Only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.