Learn More
Macherey-Nagel™ Weighing Boats, MN 808
Product Code. 10037540 Shop All Macherey-Nagel Products
Change view
Click to view available options
:
70 x 23 x 15 mm
Product Code.
10037540 70 x 23 x 15 mm
1 options
Product Code. 10037540 Supplier Macherey-Nagel™ Supplier No. 486001
Only null left
Add to basket
Add to basket
Made from a special, nitrogen-free parchment.
  • Weighing boats MN 808 are made from a special, nitrogen-free parchment
  • Used to weigh viscous or syrupy substances
Specifications
Material Nitrogen-free Parchment
Quantity 100/Pk.
Type Weighing Boat
Dimensions (L x W) 70 x 23 x 15 mm