missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WEE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | WEE2 |
|---|---|
| Concentration | 0.47 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16876575
|
Proteintech
55119-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16866575
|
Proteintech
55119-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to WEE2.
Oocyte-specific protein tyrosine kinase that phosphorylates and inhibits CDK1 and acts as a key regulator of meiosis during both prophase I and metaphase II. Required to maintain meiotic arrest in oocytes during the germinal vesicle (GV) stage, a long period of quiescence at dictyate prophase I, by phosphorylating CDK1 at ′Tyr-15′, leading to inhibit CDK1 activity and prevent meiotic reentry. Also required for metaphase II exit during egg activation by phosphorylating CDK1 at ′Tyr-15′, to ensure exit from meiosis in oocytes and promote pronuclear formation.Specifications
| WEE2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P0C1S8 | |
| 494551 | |
| Peptide | |
| Primary | |
| -20°C | |
| WEE2 |
| 0.47 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| WEE1 homolog 2 (S. pombe), Wee1 like protein kinase 1B, Wee1 like protein kinase 2, WEE1B, Wee1B kinase, WEE2 | |
| WEE2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title