Learn More
WEE1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29474-1-AP-150UL
This item is not returnable.
View return policy
Description
The WEE1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WEE1. This antibody recognizes human, mouse antigen. The WEE1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| WEE1 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Western Blot | |
| Unconjugated | |
| 150 μL | |
| 7465 | |
| IgG |