missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WEE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529303
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, NCI-H929, Neuro2A, C8D30, C2C12. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (84%), Chicken (88%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
WEE1, which is classified as a Ser/Thr kinase by sequence analysis, has been shown to inactivate multiple CDKs by tyrosine phosphorylation. Wee1 protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase.
Specifications
| WEE1 | |
| Polyclonal | |
| Unconjugated | |
| WEE1 | |
| CTRCT30; DKFZp686I18166; epididymis luminal protein 113; fa20h03; FLJ16446; HEL113; VIM; Vimentin; wee 1 homolog; WEE 1 homolog 1 (S. pombe); Wee1; WEE1 G2 checkpoint kinase; WEE1 homolog; wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; wee1A kinase; WEE1hu; wee1-like protein kinase; wu:fa20h03; zgc:136514 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22390, 7465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.45 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P30291, P47810 | |
| WEE1 | |
| Recombinant protein encompassing a sequence within the center region of human WEE1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction