missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gel Kits, 4–20%, 1.0 mm, WedgeWell™ format

Product Code. 31781177
Change view
Click to view available options
Wells:
10-well
12-well
15-well
Quantity:
10 Gels (1 Box)
2 Gels (1 Box)
30 Gels (3 Boxes)
Unit Size:
Pack of 10
Pack of 2
Pack of 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Quantity Unit Size
31781177 15-well 30 Gels (3 Boxes) Pack of 3
15456814 10-well 10 Gels (1 Box) Pack of 10
31781146 10-well 30 Gels (3 Boxes) Pack of 3
15492485 10-well 2 Gels (1 Box) Pack of 2
15466814 12-well 10 Gels (1 Box) Pack of 10
31781118 12-well 30 Gels (3 Boxes) Pack of 3
15466694 15-well 10 Gels (1 Box) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31781177 Supplier Invitrogen™ Supplier No. XP04205BOXPR
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

A25977 Mini Gel Tank and 3 x XP04205BOX Gels

Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity. Novex Tris-Glycine Mini Gels are based on traditional Laemmli protein electrophoresis that allow the use of Laemmli sample and running buffers.

Features of Novex Tris-Glycine gels, WedgeWell format, include:

  • Improved shelf life—store gels for up to 7 months at 4°C
  • Diversity—use for native and denaturing protein assays
  • Wedge-shaped wells—easily load up to two times more sample volume
  • Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments

Novex Tris-Glycine gels, WedgeWell format, come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can select from our many well formats, including 10-, 12-, and 15-well, and different pack sizes.

Run your proteins in native or denatured form

Novex Tris-Glycine gels, WedgeWell format, do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26755). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).

Convenient to use

Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) when preforming traditional wet transfer using the Invitrogen Mini Blot Module (B1000). Transfers can also be performed with rapid semi-dry transfer using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Gel Percentage 4 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine Plus
Sample Loading Volume Up to 60 μL
Separation of Protein
Separation Range 20 to 200 kDa
Separation Type Denaturing
Wells 15-well
Mode of Separation Molecular Weight
Quantity 30 Gels (3 Boxes)
Shipping Condition Room Temperature
Storage Requirements Store at 4°C to 25°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line Novex, WedgeWell
Product Type Protein Gel
For Use With (Equipment) Tetra Electrophoresis Cores
Show More Show Less
Is it okay to use protein gels past their expiration date?

We do not recommend using gels past their expiration date because over time, the polyacrylamide hydrolyzes to acrylic acid and ammonia and this will affect the resolution of the proteins. Breakdown of polyacrylamide matrix is identified by:
- Ghost bands and doublets, seen first in the high molecular weight proteins
- Smiling of dye front across the gel, with bands in outer lanes becoming very slanted - proteins run slower there due to change in pH and pore size over time.

Can I run your protein gels overnight?

This is not really recommended, but it is always possible to increase run time by lowering the voltage of the run. In general, the relationships are linear - i.e., decreasing voltage by half will double the run time.

Do I need to increase the voltage when I run a 1.5 mm protein gel versus a 1.0 mm gel?

If you are running the gel at constant voltage, you do not need to increase the voltage regardless of the thickness of the gel.

Why do you recommend running your protein gels at constant voltage?

Using constant voltage allows the current and power to decrease during the run, providing a safety margin in case of a break in the system. Having lower power is a safety benefit and will also decrease the chances of experiencing an overheating of the gel. Further, the constant voltage setting does not need adjustment to account for differences in number or thickness of gels being run.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.