Learn More
Sartorius™ Wedge for Windows
Product Code. 15459783 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Sartorius Devices
Unit Size:
Each
Product Code. 15459783
Brand: Sartorius™ YSW02
Only null left
Add to basket
Add to basket
Designed to capture data from Sartorius devices into Windows applications, specifically Microsoft Excel. Sartorius™ Wedge for Windows connects to most serial or USB devices and TCPWedge connects to most networked devices.
  • Eliminate manual data entry errors
  • Log data from multiple devices simultaneously
  • Capture real-time data to any Windows application
  • Parse and filter the data to match your requirements
  • 100% accurate, reliable, and compliant
TRUSTED_SUSTAINABILITY
Specifications
Includes WinWedge and TCPWedge capture programs
For Use With (Application) Windows applications, Microsoft Excel
For Use With (Equipment) Sartorius Devices