Learn More
Thermo Scientific™ WebSeal™ 96-Well Mats for WebSeal Glass Inserted Plate Kits
Product Code. 15152789
Click to view available options
Pre-slit:
No
Yes
Product Type:
WebSeal Mat, Solid Plugs
WebSeal Mat, Welled Plugs
Unit Size:
Each
Product Code. 15152789
Brand: Thermo Scientific™ 60180M157
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Extend the usefulness of your well plate kits with replacement mats.
Extend the usefulness of your wellplate kits with WebSeal 96-Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
Extend the usefulness of your wellplate kits with WebSeal 96-Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone, PTFE
No. of Wells 96
No. per Pack 1 Mat
For Use With (Application) Sample Storage
For Use With (Equipment) WebSeal Glass Inserted Plate Kit (Cat. No. 60180-K109)
Pre-slit No
Product Type WebSeal Mat, Solid Plugs
Product Line Webseal
Plate Compatibility WebSeal Glass Inserted 96-well Plate
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.