missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR90 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121498
This item is not returnable.
View return policy
Description
Required for efficient primary cilium formation.
Specifications
| WDR90 | |
| Polyclonal | |
| Unconjugated | |
| WDR90 | |
| C16orf15; C16orf16; C16orf17; C16orf18; C16orf19; KIAA1924; WD repeat domain 90; WD repeat-containing protein 90; WDR90 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100519302, 102555298, 197335 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96KV7 | |
| WDR90 | |
| Met1-Pro235 (Accession Q96KV7), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction