missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR83OS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566788
This item is not returnable.
View return policy
Description
Immunogen sequence: MSTNNMSDPRR PNKVLRYKPP PSECNPA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
WDR83OS is a protein coding gene.
Specifications
| WDR83OS | |
| Polyclonal | |
| Unconjugated | |
| WDR83OS | |
| C19orf56; CGI-140; My006; Protein Asterix; PTD008; UPF0139 membrane protein C19orf56; WD repeat domain 83 opposite strand; WDR83 opposite strand; WDR83OS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51398 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y284 | |
| WDR83OS | |
| Recombinant Human WDR83OS. Recombinant protein control fragment (Product #RP-105363). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction