missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR83 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545085
This item is not returnable.
View return policy
Description
Peptide sequence: EGALALALPV GSGVVQSLAY HPTEPCLLTA MGGSVQCWRE EAYEAEDGAG Sequence homology: Cow: 85%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 85%; Rabbit: 79%; Rat: 85%.
MORG1 is a member of the WD-40 protein family. The protein is proposed to function as a molecular scaffold for various multimeric protein complexes. The protein associates with several components of the extracellular signal-regulated kinase (ERK) pathway, and promotes ERK activity in response to serum or other signals. The protein also interacts with egl nine homolog 3 (EGLN3, also known as PHD3) and regulates expression of hypoxia-inducible factor 1, and has been purified as part of the spliceosome.
Specifications
| WDR83 | |
| Polyclonal | |
| Unconjugated | |
| WDR83 | |
| 1500041N16Rik; MAPK cascade scaffold protein; MAPK organizer 1; MGC4238; Mitogen-activated protein kinase organizer 1; MORG1; RGD1306947; WD repeat domain 83; WD repeat domain containing 83; WD repeat domain-containing protein 83; WD repeat protein Morg1; WDR83 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84292 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BRX9 | |
| WDR83 | |
| Synthetic peptide directed towards the C-terminal of human WDR83 (aa 266-315). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction