missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR82 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA549392

Product Code. 15917384

  • £441.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Predicted to react with xenopus and chicken based on sequence homology.

WDR82 is a component of the mammalian SET1A (MIM 611052)/SET1B (MIM 611055) histone H3-Lys4 methyltransferase complexes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WDR82
Polyclonal
Unconjugated
WDR82
9430077D24Rik; Cdw5; CDW5/WDR82; MST107; MSTP107; PRO2730; PRO34047; Protein TMEM113; Protein word of mouth; PTK9L; Swd2; TMEM113; transmembrane protein 113; UNQ9342/PRO34047; WD repeat domain 82; WD repeat domain containing 82; WD repeat-containing protein 82; Wdr82; wdr82 protein; WDR82A; wom; word of mouth; wu:fb22h06; zgc:55453; zgc:85635; zgc:91817
Rabbit
Antigen affinity chromatography, Protein A
RUO
321614, 77305, 80335
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6NV31, Q6UXN9, Q8BFQ4
WDR82
KLH conjugated synthetic peptide between 50-85 amino acids from the N-terminal region of human WDR82.
400 μL
Primary
Human, Mouse, Zebrafish
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.