missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR82 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549393
This item is not returnable.
View return policy
Description
Predicted to react with mouse based on sequence homology.
WDR82 is a component of the mammalian SET1A (MIM 611052)/SET1B (MIM 611055) histone H3-Lys4 methyltransferase complexes.
Specifications
| WDR82 | |
| Polyclonal | |
| Unconjugated | |
| WDR82 | |
| 9430077D24Rik; Cdw5; CDW5/WDR82; MST107; MSTP107; PRO2730; PRO34047; Protein TMEM113; Protein word of mouth; PTK9L; Swd2; TMEM113; transmembrane protein 113; UNQ9342/PRO34047; WD repeat domain 82; WD repeat domain containing 82; WD repeat-containing protein 82; Wdr82; wdr82 protein; WDR82A; wom; word of mouth; wu:fb22h06; zgc:55453; zgc:85635; zgc:91817 | |
| Rabbit | |
| Affinity Chromatography, Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 77305, 80335 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6UXN9, Q8BFQ4 | |
| WDR82 | |
| KLH conjugated synthetic peptide between 12~46 amino acids from the N-terminal region of human WDR82. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction