missing translation for 'onlineSavingsMsg'
Learn More

WDR77 Rabbit anti-Human, Polyclonal, Invitrogen™

Product Code. 17482013 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
17482013 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 17482013

Brand: Thermo Scientific PA5118360

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WDR77 Polyclonal antibody specifically detects WDR77 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR77
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% Proclin 300; pH 7
Gene WDR77
Gene Accession No. Q9BQA1
Gene Alias 2610003I18Rik; 2610312E17Rik; Ac2-269; androgen receptor cofactor p44; C79984; HKMT1069; MEP50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77
Host Species Rabbit
Immunogen E. coli-derived Human WDR77 fragment
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79084
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.