missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR74 Antibody (OTI4B2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74867CL1
This item is not returnable.
View return policy
Description
WDR74 Monoclonal antibody specifically detects WDR74 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| WDR74 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ10439, FLJ21730, NOP seven-associated protein 1, NSA1, WD repeat domain 74, WD repeat-containing protein 74 | |
| Human recombinant protein fragment corresponding to amino acids 114-385 of human WDR74 (NP_060563) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 54663 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI4B2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction