missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR66 Antibody (2A6F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37381-0.025ml
This item is not returnable.
View return policy
Description
WDR66 Monoclonal specifically detects WDR66 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| WDR66 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| FLJ39783, MGC33630, WD repeat domain 66, WD repeat-containing protein 66 | |
| Mouse | |
| 130 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 2A6F7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| WDR66 | |
| Purified recombinant fragment of human WDR66 (AA: 1-250) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 144406 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction