missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR63 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.
WDR63 contains 5 WD repeats. The exact function of WDR63 is not known.
Specifications
Specifications
| Antigen | WDR63 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | WDR63 |
| Gene Accession No. | Q8IWG1 |
| Gene Alias | DIC3; NYD-SP29; testicular tissue protein Li 225; Testis development protein NYD-SP29; testis development protein NYD-SP29 (NYD-SP29); WD repeat domain 63; WD repeat-containing protein 63; WDR63 |
| Gene Symbols | WDR63 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?