missing translation for 'onlineSavingsMsg'
Learn More

WDR6 Antibody (2D4), Novus Biologicals™

Product Code. 18315839 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18315839 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18315839 Supplier Novus Biologicals Supplier No. H00011180M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WDR6 Monoclonal antibody specifically detects WDR6 in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH02826
Gene Alias FLJ10218, FLJ52552, FLJ56107, MGC126756, MGC142027, WD repeat domain 6, WD repeat-containing protein 6
Host Species Mouse
Immunogen WDR6 (AAH02826, 1 a.a. ~ 289 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MHLSSHRLDEYWDRQRNRHRMVKVDPETRYMSLAVCELDQPGLGPLVAAACSDGAVRLFLLQDSGRILQLLAETFHHKRCVLKVHSFTHEAPNQRRRLLLCSAATDGSLAFWDLTTMLDHDSTVLEPPVDPGLPYRLGTPSLTLQAHSCGINSLHTLPTREGHHLVASGSEDGSLHVFVLAVEMLQLEEAVGEAGLVPQLRVLEEYSVPCAHAAHVTGLKILSPSIMVSASIDQRLTFWRLGHGEPTFMNSTVFHVPDVADMDCWPVSPEFGHRCALGGQGLEVYNWYD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11180
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.