missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR59 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565506
This item is not returnable.
View return policy
Description
Immunogen sequence: GSLTYSDPRER ERDQHDKNKR LLDPANTQQF DDFKKCYGEI LYRWGLREKR AEVLKFVSCP PDPHKGIEFG VYCSHCRSEV RGTQCAICKG FTFQCAIC Highest antigen sequence identity to the following orthologs - mouse 100%, rat 71%.
As a component of the GATOR subcomplex GATOR2, functions within the amino acid-sensing branch of the TORC1 signaling pathway. Indirectly activates mTORC1 and the TORC1 signaling pathway through the inhibition of the GATOR1 subcomplex (PubMed:23723238). It is negatively regulated by the upstream amino acid sensors SESN2 and CASTOR1 (PubMed:25457612, PubMed:27487210). [UniProt].
Specifications
| WDR59 | |
| Polyclonal | |
| Unconjugated | |
| WDR59 | |
| FP977; GATOR complex protein WDR59; KIAA1923; WD repeat domain 59; WD repeat-containing protein 59; WDR59 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PJI9 | |
| WDR59 | |
| Recombinant Human WDR59. Recombinant protein control fragment (Product #RP-104851). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction