missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR59 Monoclonal Antibody (K16273_16H3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556211
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: RKQKEGSKDY QLVTWSRDQT LRMWRVDSQM QRLCANDILD GVDEFIESIS LLPEPEKTLH TEDTDHQHTA SHGEEEALKE DPPRNLLEER KSDQLGLPQT
As a component of the GATOR subcomplex GATOR2, functions within the amino acid-sensing branch of the TORC1 signaling pathway. Indirectly activates mTORC1 and the TORC1 signaling pathway through the inhibition of the GATOR1 subcomplex (PubMed:23723238). It is negatively regulated by the upstream amino acid sensors SESN2 and CASTOR1 (PubMed:25457612, PubMed:27487210). [UniProt].
Specifications
| WDR59 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q6PJI9 | |
| WDR59 | |
| Recombinant human WDR59 (immunogen range is 291-390). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K16273_16H3 | |
| Unconjugated | |
| WDR59 | |
| FP977; GATOR complex protein WDR59; KIAA1923; WD repeat domain 59; WD repeat-containing protein 59; WDR59 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction