missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR59 Monoclonal Antibody (K16273_16H3), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA555831

 View more versions of this product

Product Code. 30205315

  • £380.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: RKQKEGSKDY QLVTWSRDQT LRMWRVDSQM QRLCANDILD GVDEFIESIS LLPEPEKTLH TEDTDHQHTA SHGEEEALKE DPPRNLLEER KSDQLGLPQT

As a component of the GATOR subcomplex GATOR2, functions within the amino acid-sensing branch of the TORC1 signaling pathway. Indirectly activates mTORC1 and the TORC1 signaling pathway through the inhibition of the GATOR1 subcomplex (PubMed:23723238). It is negatively regulated by the upstream amino acid sensors SESN2 and CASTOR1 (PubMed:25457612, PubMed:27487210). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WDR59
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q6PJI9
WDR59
Recombinant human WDR59 (immunogen range is 291-390).
50 μL
Primary
Human
Antibody
IgG2a
Western Blot
K16273_16H3
Unconjugated
WDR59
FP977; GATOR complex protein WDR59; KIAA1923; WD repeat domain 59; WD repeat-containing protein 59; WDR59
Mouse
Protein A
RUO
79726
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.