missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR58 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94392-0.1ml
This item is not returnable.
View return policy
Description
WDR58 Polyclonal antibody specifically detects WDR58 in Human, Mouse samples. It is validated for Western Blot
Specifications
| WDR58 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| fSAP35WDR58, Functional spliceosome-associated protein 35, MGC2655, THO complex 6 homolog (Drosophila), THO complex subunit 6 homolog, WD repeat domain 58, WD repeat-containing protein 58 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human WDR58 (NP_077315.2). IFSLSSALSSEAKEESKKPVVTFQAHDGPVYSMVSTDRHLLSAGDGEVKAWLWAEMLKKGCKELWRRQPPYRTSLEVPEINALLLVPKENSLILAGGDCQL | |
| 0.1 mL | |
| Protein Kinase | |
| 79228 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction