missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR54 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560298
This item is not returnable.
View return policy
Description
Immunogen sequence: RVLVFDIPAK GPNIVLSEEL AGHQMPITDI ATEPAQGQDC VADMVTADDS GLLCVWRSGP EFTLLTRIPG FGVPCPS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 91%.
Specifications
| WDR54 | |
| Polyclonal | |
| Unconjugated | |
| WDR54 | |
| 1700030E05Rik; D3Mm3e; WD repeat domain 54; WD repeat-containing protein 54; WDR54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84058 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H977 | |
| WDR54 | |
| Recombinant protein corresponding to Human WDR54. Recombinant protein control fragment (Product #RP-99634). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction