missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR53 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553415
This item is not returnable.
View return policy
Description
Immunogen sequence: PVLCLNASKE GLLASGAEGG DLTAWGEDGT PLGHTRFQGA DDVTSVLFSP SCPTKLYASH GETISVLDVR SLKDSLDHFH VNEEEINCLS LNQTENLLAS ADDSGAIKIL DLENKKVIRS LKRHSNICSS V Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.
Specifications
| WDR53 | |
| Polyclonal | |
| Unconjugated | |
| WDR53 | |
| 1500002B03Rik; AI848860; RGD1559546; WD domain, G-beta repeat-containing protein; WD repeat domain 53; WD repeat-containing protein 53; WDR53 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 348793, 498097, 68980 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z5U6, Q9DB94 | |
| WDR53 | |
| Recombinant protein corresponding to Human WDR53. Recombinant protein control fragment (Product #RP-90778). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction