missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ WDR51A Recombinant Protein
£280.00 - £425.00
Specifications
Accession Number | AAH07417 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 25886 |
Molecular Weight (g/mol) | 65.78 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16177852
|
Abnova™
H00025886-P01.10UG |
10 ug |
£280.00
10µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16187852
|
Abnova™
H00025886-P01.25UG |
25 ug |
£425.00
25µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH07417 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
65.78 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WDR51A | |
Human | |
Recombinant | |
Solution |
Antibody Production, ELISA, Protein Array, Western Blot | |
25886 | |
WDR51A (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVWHMKPQSRAYRFTGHKDAVTCVNFSPSGHLLASGSRDKTVRIWVPNVKGESTVFRAHTATVRSVHFCSDGQSFVTASDDKTVKVWATHRQKFLFSLSQHINWVRCAKFSPDGRLIVSASDDKTVKLWDKSSRECVHSYCEHGGFVTYVDFHPSGTCIAAAGMDNTVKVWDVRTHRLLQHYQLHSAAVNGLSFHPSGNYLITASSDSTLKILDLMEGRLLYTLHGHQGPATTVAFSRTGEYFASGGSDEQVMVWKSNFDIVDHGEVTKVPRPPATLASSMGNLPEVDFPVPPGRGRSVESVQSQPQEPVSVPQTLTSTLEHIVGQLDVLTQTVSILEQRLTLTEDKLKQCLENQQLIMQRATP | |
DKFZp434C245/MGC131902 | |
WDR51A | |
Wheat Germ (in vitro) | |
GST |