missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals WDR51A Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00025886-B01P
This item is not returnable.
View return policy
Description
WDR51A Polyclonal antibody specifically detects WDR51A in Human samples. It is validated for Western Blot
Specifications
| WDR51A | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH07417 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| DKFZp434C245, MGC131902, PIX2, POC1 centriolar protein homolog A, POC1 centriolar protein homolog A (Chlamydomonas), WD repeat domain 51A, WD repeat-containing protein 51A, WDR51A | |
| WDR51A (AAH07417, 1 a.a. - 364 a.a.) full-length human protein. MVWHMKPQSRAYRFTGHKDAVTCVNFSPSGHLLASGSRDKTVRIWVPNVKGESTVFRAHTATVRSVHFCSDGQSFVTASDDKTVKVWATHRQKFLFSLSQHINWVRCAKFSPDGRLIVSASDDKTVKLWDKSSRECVHSYCEHGGFVTYVDFHPSGTCIAAAGMDNTVKVWDVRTHRLLQHYQLHSAAVNGLSFHPSGNYLITASSDSTLKILDLMEGRLLYTLHGHQGPATTVAFSRTGEYFASGGSDEQVMVWKSNFDIVDHGEVTKVPRPPATLASSMGNLPEVDFPVPPGRGRSVESVQSQPQEPVSVPQTLTSTLEHIVGQLDVLTQTVSILEQRLTLTEDKLKQCLENQQLIMQRATP | |
| 0.05 mg | |
| Cell Biology, Cell Cycle and Replication | |
| 25886 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction