missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | WDR5 |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:100-1:500 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18333555
|
Novus Biologicals
NBP3-18672-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18328846
|
Novus Biologicals
NBP3-18672-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WDR5 Polyclonal antibody specifically detects WDR5 in Mouse samples. It is validated for Western Blot, ELISASpecifications
| WDR5 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers | |
| Whole antiserum | |
| BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5 | |
| Polyclonal antibody raised in rabbit against mouse Wdr5 (WD (tryptophan-aspartate) repeat domain protein 5), using two KLH-conjugated synthetic peptides containing an amino acid sequence from the central part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:100-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse | |
| P61964 | |
| 11091 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title