missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117743
This item is not returnable.
View return policy
Description
WDR5 Polyclonal Antibody for Western Blot, ICC/IF, ChIP, ChIP-seq
Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.
Specifications
| WDR5 | |
| Polyclonal | |
| Unconjugated | |
| WDR5 | |
| 2410008O07Rik; AA408785; AA960360; Big; BIG3; BIG-3; BMP2-induced 3-kb gene protein; CFAP89; cilia and flagella associated protein 89; SWD3; SWD3, Set1c WD40 repeat protein, homolog; Unknown (protein for MGC:137327); WD repeat domain 5; WD repeat-containing protein 5; WD repeat-containing protein BIG-3; Wdr5; wu:fk47f04; zgc:56591; zgc:76895; zgc:76895 protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11091 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 0.05% ProClin 300, 0.05% sodium azide | |
| P61964 | |
| WDR5 | |
| WDR5 recombinant protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction