missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR5 Antibody (RAB-C223), Alexa Fluor™ 405 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20133AF405
This item is not returnable.
View return policy
Description
WDR5 Monoclonal antibody specifically detects WDR5 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| WDR5 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing WDR5 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 11091 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| RAB-C223 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction