missing translation for 'onlineSavingsMsg'
Learn More

WDR5 Antibody, Novus Biologicals™

Product Code. 18325073
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325073 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325073 Supplier Bio-Techne Supplier No. NBP318669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WDR5 Polyclonal antibody specifically detects WDR5 in Human samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP), ChIP assay Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR5
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Chromatin Immunoprecipitation 2 ug/ChIP, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:2500 - 1:5000, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing
Formulation PBS
Gene Accession No. P61964
Gene Alias BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human WDR5 (WD (tryptophan-aspartate) repeat domain 5), using a recombinant protein.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.