missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR5 Antibody (18D11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26253-100ul
This item is not returnable.
View return policy
Description
WDR5 Monoclonal antibody specifically detects WDR5 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| WDR5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 18D11 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5 | |
| A synthesized peptide derived from Human WDR5 [UniProt P61964] | |
| 100 μL | |
| Chromatin Modifiers | |
| 11091 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction