missing translation for 'onlineSavingsMsg'
Learn More

WDR5-0103, MedChemExpress

Product Code. 16443717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443717 50 mg 50mg
16430208 5 mg 5mg
16417606 100 mg 100mg
16407748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443717 Supplier Medchem Express Supplier No. HY19347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WDR5-0103 is a potent and selective WD repeat-containing protein 5 (WDR5) antagonist with Kd of 450 nM.

WDR5-0103 is a potent and selective WD repeat-containing protein 5 (WDR5) antagonist with Kd of 450 nM.

Specifications

Chemical Name or Material WDR5-0103
CAS 890190-22-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H25N3O4
Quantity 50 mg
Synonym WD-Repeat Protein 5-0103
Solubility Information DMSO : 50 mg/mL (130.40 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(N2CCN(C)CC2)C(NC(C3=CC=CC(OC)=C3)=O)=C1
Molecular Weight (g/mol) 383.44
Formula Weight 383.44
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.