missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ WDR46 Recombinant Protein Antigen
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.1mL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human WDR46. The WDR46 Recombinant Protein Antigen is derived from E. coli. The WDR46 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-94128. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Spécification
Spécification
| Gene ID (Entrez) | 9277 |
| Species | Human |
| Purification Method | Chromatography |
| Purity | >80% |
| Concentration | 0.5mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Symbol | WDR46 |
| Label Type | Unlabeled |
| Afficher plus |
For Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu