missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR45 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19194-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene has a pseudogene at chromosome 4q31.3. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity and full-length nature of some variants have not been determined.Specifications
| WDR45 | |
| Polyclonal | |
| Unconjugated | |
| WDR45 | |
| JM5, WD repeat domain 45, WDR45, WDRX1, WDRXI4, WIPI 4, WIPI4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11152 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y484 | |
| WDR45 | |
| WDR45 Fusion Protein Ag6741 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction