missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR45 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577801
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of PA5-77801 was sufficient for detection of WIPI4 in 15 μg of Human HeLa Cell lysates by ECL immunoblot analysis using goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 40 kDa.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene has a pseudogene at chromosome 4q31.3. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity and full-length nature of some variants have not been determined.
Specifications
| WDR45 | |
| Polyclonal | |
| Unconjugated | |
| WDR45 | |
| JM5; NBIA4; NBIA5; neurodegeneration with brain iron accumulation 4; neurodegeneration with brain iron accumulation 5; WD repeat domain 45; WD repeat domain phosphoinositide-interacting protein 4; WD repeat domain, X-linked 1; WD repeat-containing protein 45; WD45 repeat protein interacting with phosphoinositides 4; WDR45; WDRX1; WDRXI4; WIPI4; WIPI-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11152, 302559 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| Q5U2Y0, Q9Y484 | |
| WDR45 | |
| Synthetic peptide from the C-terminal of Human WIPI4 (aa 312-322). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction