missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR45 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18255F
This item is not returnable.
View return policy
Description
WDR45 Polyclonal antibody specifically detects WDR45 in Human, Rat samples. It is validated for Immunofluorescence
Specifications
| WDR45 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| JM5, WD repeat domain 45, WD repeat domain phosphoinositide-interacting protein 4, WD repeat-containing protein 45, WD45 repeat protein interacting with phosphoinositides 4, WDRX1, WDRXI4, WIPI-4, X-linked 1 | |
| Synthetic peptide from the C-terminal of Human WDR45(aa. 312-322) | |
| 0.1 mL | |
| Autophagy, Cell Biology | |
| 11152 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction