missing translation for 'onlineSavingsMsg'
Learn More

WDR4 Antibody (1F9), Novus Biologicals™

Product Code. 18321009 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321009 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321009 Supplier Novus Biologicals Supplier No. H00010785M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WDR4 Monoclonal antibody specifically detects WDR4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1F9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01074
Gene Alias tRNA (guanine-N(7)-)-methyltransferase subunit WDR4, WD repeat domain 4, 10TRM82, WD repeat-containing protein 4
Host Species Mouse
Immunogen WDR4 (AAH01074, 1 a.a. ~ 266 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLLDVAVSPDDRFILTADRDEKIRVSWAAAPHSIESFCLGHTEFVSRISVVPTQPGLLLSSSGDGTLRLWEYRSGRQLHCCHLASLQELVDPQAPQKFAASRIAFWCQENCVALLCDGTSVVYIFQLDARRQQLVYRQQLAFQHQVWDVAFEETQGLWVLQDCQEAPLVLYRPVGDQWQSVPESTVLKKVSGVLRGNWAMLEGSAGADASFSSLYKATFDNVTSYLKKKEERLQQQLEKKQRRRSPPPGPDGHAKKMRPGEATLSC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10785
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.