missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558110
This item is not returnable.
View return policy
Description
Immunogen sequence: APLNVSMSPT GDFLATSHVD HLGIYLWSNI SLYSVVSLRP LPADYVPSIV MLPGTCQTQD VEVSEETVEP SDELIEYDSP EQLNEQLVTL S Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Mutations in this gene have been associated with adult-onset primary open-angle glaucoma (POAG).
Specifications
| WDR36 | |
| Polyclonal | |
| Unconjugated | |
| WDR36 | |
| GLC1G; TAWDRP; TA-WDRP; T-cell activation WD repeat protein; T-cell activation WD repeat-containing protein; UTP21; WD repeat domain 36; WD repeat-containing protein 36; WDR36 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 134430 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NI36 | |
| WDR36 | |
| Recombinant protein corresponding to Human WDR36. Recombinant protein control fragment (Product #RP-102785). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction