missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR36 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93299-0.1ml
This item is not returnable.
View return policy
Description
WDR36 Polyclonal antibody specifically detects WDR36 in Human samples. It is validated for Western Blot
Specifications
| WDR36 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| DKFZp686I1650, TA-WDRPopen angle, G, WD repeat domain 36, WD repeat-containing protein 36 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 702-951 of human WDR36 (NP_644810.1). LRPLPADYVPSIVMLPGTCQTQDVEVSEETVEPSDELIEYDSPEQLNEQLVTLSLLPESRWKNLLNLDVIKKKNKPKEPPKVPKSAPFFIPTIPGLVPRYAAPEQNNDPQQSKVVNLGVLAQKSDFCLKLEEGLVNNKYDTALNLLKESGPSGIETELRSLSPDCGGSIEVMQSFLKMIGMMLDRKRDFELAQAYLALFLKLHLKMLPSEPVLLEEITNLSSQVEENWTHLQSLFNQSMCILNYLKSALL | |
| 0.1 mL | |
| Vision | |
| 134430 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction