missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
WDR35 Polyclonal antibody specifically detects WDR35 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Gene Alias | CED2, IFT121, WD repeat domain 35 |
| Immunogen | WDR35 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human WDR35. The immunogen is located within amino acids 60 - 110 of WDR35. Amino Acid Squence: TWNEQYQKLTTSDENG |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?