missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565445
This item is not returnable.
View return policy
Description
Immunogen sequence: LAWHPLGHILC SGSNDHTSKF WTRNRPGDKM RDRYNLNLLP GMSEDGVEYD DLEPNSLAVI PGMGIPEQLK LAMEQEQMGK DESNEIEMTI PGLDWGMEEV MQKDQKKVPQ KKVPYAKPIP AQFQQAWMQN KVPIP Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
WDR33 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| WDR33 | |
| Polyclonal | |
| Unconjugated | |
| Wdr33 | |
| 1110001N06Rik; 2310011G05Rik; 2810021O11Rik; 8430413N20Rik; FLJ11294; FLJ11294-like; NET14; pre-mRNA 3' end processing protein WDR33; WD repeat domain 33; WD repeat-containing protein 33; WD repeat-containing protein of 146 kDa; WD repeat-containing protein WDC146; WDC146; Wdr33; WD-repeat protein 33; zgc:110570 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55339 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0J8 | |
| Wdr33 | |
| Recombinant Human WDR33. Recombinant protein control fragment (Product #RP-106125). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction