missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554998
This item is not returnable.
View return policy
Description
Immunogen sequence: GARLFGWLKE RSLGRGLFVD PARDNFRTMT SLYGSIHPAD SVYLSTRTHG AVFNLEYSPD GSVLTVACEQ TEVLL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
DCAF10 may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
Specifications
| WDR32 | |
| Polyclonal | |
| Unconjugated | |
| DCAF10 | |
| AA959934; DCAF10; DDB1 and CUL4 associated factor 10; DDB1- and CUL4-associated factor 10; RGD1304730; WD repeat domain 32; WD repeat-containing protein 32; WDR32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79269 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5QP82 | |
| DCAF10 | |
| Recombinant protein corresponding to Human WDR32. Recombinant protein control fragment (Product #RP-92941). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction