missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR23 Polyclonal Antibody

WDR23 Polyclonal Antibody
Brand: Thermo Scientific PA556658
This item is not returnable.
View return policy
Description
WDR23 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| WDR23 | |
| Polyclonal | |
| Unconjugated | |
| Dcaf11 | |
| 0710008A13Rik; C76035; D14Ucla1; Dacf11; DCAF11; DDB1 and CUL4 associated factor 11; DDB1- and CUL4-associated factor 11; DKFZp779A1629; GL014; GLO14; PRO2389; WD repeat domain 23; WD repeat domain 23 isoform 1; WD repeat-containing protein 23; WDR23; WDR23 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEB1 | |
| Dcaf11 | |
| Recombinant protein corresponding to Human WDR23. Recombinant protein control fragment (Product #RP-89040). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction