missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR21C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564329
This item is not returnable.
View return policy
Description
Immunogen sequence: AGGSKYGIIT MRGLTTPELR VYPHK Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.
Specifications
| WDR21C | |
| Polyclonal | |
| Unconjugated | |
| DCAF4L2 | |
| DCAF4L2; DDB1 and CUL4 associated factor 4 like 2; DDB1 and CUL4 associated factor 4-like 2; DDB1- and CUL4-associated factor 4-like protein 2; WD repeat domain 21C; WD repeat-containing protein 21C; WDR21C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 138009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NA75 | |
| DCAF4L2 | |
| Recombinant protein corresponding to Human WDR21C. Recombinant protein control fragment (Product #RP-103657). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction