missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR16 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12644-1-AP-150UL
This item is not returnable.
View return policy
Description
WD repeat-containing proteins, such as WDR16, play crucial roles in a wide range of physiologic functions, including signal transduction, RNA processing, remodeling the cytoskeleton, regulation of vesicular traffic, and cell division (Silva et al., 2005 ).Specifications
| WDR16 | |
| Polyclonal | |
| Unconjugated | |
| CFAP52 | |
| WD repeat domain 16, WDR16, WDRPUH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146845 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N1V2 | |
| CFAP52 | |
| WDR16 Fusion Protein Ag3281 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction