missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541985
This item is not returnable.
View return policy
Description
Peptide sequence: DTRSCKAPLY DLAAHEDKVL SVDWTDTGLL LSGGADNKLY SYRYSPTTSH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 89%.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein is highly similar to the mouse WD repeat domain 12 protein at the amino acid level. The protein encoded by this gene is a component of a nucleolar protein complex that affects maturation of the large ribosomal subunit.
Specifications
| WDR12 | |
| Polyclonal | |
| Unconjugated | |
| WDR12 | |
| 4933402C23Rik; fb24f09; HAMAP-Rule:MF_03029}; MNCb-5414; nuclear protein Ytm1; ribosome biogenesis protein wdr12; ribosome biogenesis protein WDR12 {ECO:0000255; WD repeat domain 12; WD repeat domain 12 protein; WD repeat-containing protein 12; WD repeat-containing protein 12 {ECO:0000255; WDR12; wdr12 protein; wu:fb24f09; YTM1; Ytm1p; zgc:55609 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55759 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9GZL7 | |
| WDR12 | |
| Synthetic peptide directed towards the C-terminal of human WDR12 (aa 371-420). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction