missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
WDR12 Polyclonal specifically detects WDR12 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | WDR12 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | FLJ10881, FLJ12719, FLJ12720, ribosome biogenesis protein WDR12, WD repeat domain 12, WD repeat-containing protein 12, YTM1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human WDR12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?