missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13676-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| WDR1 | |
| Polyclonal | |
| Unconjugated | |
| WDR1 | |
| Actin interacting protein 1, AIP1, NORI 1, WD repeat containing protein 1, WD repeat domain 1, WDR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22388, 360950, 9948 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75083, O88342, Q5RKI0 | |
| Wdr1 | |
| WDR1 Fusion Protein Ag4670 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction