Learn More
WDR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13676-1-AP-20UL
 View more versions of this product
Product Code. 16802024
  • £135.00 / 20µL
Description
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
WDR1
Polyclonal
Unconjugated
WDR1
Actin interacting protein 1, AIP1, NORI 1, WD repeat containing protein 1, WD repeat domain 1, WDR1
Rabbit
Antigen Affinity Chromatography
RUO
22388, 360950, 9948
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75083, O88342, Q5RKI0
Wdr1
WDR1 Fusion Protein Ag4670
20 μL
Primary
Rat, Mouse, Human